(4-tert-Butylphenyl)hydrazine

Supplier: Matrix Scientific

037563-500MG
101843-670EA 230.77 USD
101843-670
(4-tert-Butylphenyl)hydrazine
(4-tert-Butylphenyl)hydrazine

Hazard: Irritant

Formula: C₁₀H₁₆N₂
MDL Number: MFCD00177173
CAS Number: 61765-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR