4-Bromo-2,5-dichloro-3-thiophenesulfonyl chloride
Supplier: Matrix Scientific
037555-500MG
101843-654EA
193.85
USD
101843-654
4-Bromo-2,5-dichloro-3-thiophenesulfonyl chloride
4-Bromo-2,5-dichloro-3-thiophenesulphonyl chloride
Hazard: Irritant
Formula:
C₄BrCl₃O₂S₂ |
MDL Number:
MFCD00051664 CAS Number: 166964-36-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...