2-[(2-Aminoethyl)thio]-N-(2,6-dimethylphenyl)acetamide hydrochloride

Supplier: Matrix Scientific

037552-500MG
101843-648EA 146.15 USD
101843-648
2-[(2-Aminoethyl)thio]-N-(2,6-dimethylphenyl)acetamide hydrochloride
2-[(2-Aminoethyl)thio]-N-(2,6-dimethylphenyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₉ClN₂OS
MDL Number: MFCD00445670
CAS Number: 312926-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR