2-Amino-4-[(3,4-diaminophenyl)sulfonyl]phenylamine

Supplier: Matrix Scientific

Synonyms: 4,4'-sulphonylbis(benzene-1,2-diamine)4,4'-Sulfonylbis(benzene-1,2-diamine)

037539-500MG
101843-622EA 476.92 USD
101843-622
2-Amino-4-[(3,4-diaminophenyl)sulfonyl]phenylamine
2-Amino-4-[(3,4-diaminophenyl)sulfonyl]phenylamine

Hazard: Irritant

Formula: C₁₂H₁₄N₄O₂S
MW: 278.33 g/mol
MDL Number: MFCD00437381
CAS Number: 13224-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR