1-(2,4-Dichlorophenyl)-2-(3,5-dimethyl-1H-1,2,4-triazol-1-yl)ethanone

Supplier: Matrix Scientific

037536-500MG
101843-616EA 220.87 USD
101843-616
1-(2,4-Dichlorophenyl)-2-(3,5-dimethyl-1H-1,2,4-triazol-1-yl)ethanone
1-(2,4-Dichlorophenyl)-2-(3,5-dimethyl-1H-1,2,4-triazol-1-yl)ethanone

Hazard: Irritant

Formula: C₁₂H₁₁Cl₂N₃O
MDL Number: MFCD00804956
CAS Number: 169773-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR