3-[3,5-Di(tert-butyl)-4-hydroxyphenyl]propanohydrazide

Supplier: Matrix Scientific

037531-500MG
101843-604EA 193.85 USD
101843-604
3-[3,5-Di(tert-butyl)-4-hydroxyphenyl]propanohydrazide
3-[3,5-Di(tert-butyl)-4-hydroxyphenyl]propanohydrazide

Hazard: Irritant

Formula: C₁₇H₂₈N₂O₂
MDL Number: MFCD00411252
CAS Number: 32687-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR