1-(Bromomethyl)adamantane

Supplier: Matrix Scientific

037530-500MG
101843-602EA 289.8 USD
101843-602
1-(Bromomethyl)adamantane
1-(Bromomethyl)adamantane

Hazard: Irritant

Formula: C₁₁H₁₇Br
MDL Number: MFCD00185303
CAS Number: 14651-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR