N-[4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-N-methylpropan-2-amine

Supplier: Matrix Scientific

037524-500MG
101843-594EA 193.85 USD
101843-594
N-[4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-N-methylpropan-2-amine
N-[4-Methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]-N-methylpropan-2-amine

Hazard: Irritant

Formula: C₁₈H₃₀BNO₃
MDL Number: MFCD12027134
CAS Number: 934586-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR