2-Bromo-N-(3-chloro-2-methylphenyl)propanamide
Supplier: Matrix Scientific
037519-500MG
101843-584EA
146.15
USD
101843-584
2-Bromo-N-(3-chloro-2-methylphenyl)propanamide
2-Bromo-N-(3-chloro-2-methylphenyl)propanamide
Hazard: Irritant
Formula:
C₁₀H₁₁BrClNO |
MDL Number:
MFCD12027130 CAS Number: 127091-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...