2-Bromo-N-mesitylpropanamide

Supplier: Matrix Scientific

037517-500MG
101843-580EA 146.15 USD
101843-580
2-Bromo-N-mesitylpropanamide
2-Bromo-N-mesitylpropanamide

Hazard: Irritant

Formula: C₁₂H₁₆BrNO
MDL Number: MFCD00510153
CAS Number: 104509-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR