2-Bromo-N-(2,3-dimethylphenyl)propanamide

Supplier: Matrix Scientific

037516-500MG
101843-578EA 146.15 USD
101843-578
2-Bromo-N-(2,3-dimethylphenyl)propanamide
2-Bromo-N-(2,3-dimethylphenyl)propanamide

Hazard: Irritant

Formula: C₁₁H₁₄BrNO
MDL Number: MFCD12027129
CAS Number: 1119451-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR