Diethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate

Supplier: Matrix Scientific

037513-500MG
101843-572EA 193.85 USD
101843-572
Diethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Diethyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄S
MDL Number: MFCD01176952
CAS Number: 24264-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR