1-Thien-2-ylpropan-1-amine

Supplier: Matrix Scientific

Synonyms: 1-(Thiophen-2-yl)propan-1-amine hydrochloride

037511-500MG
101843-566EA 126.5 USD
101843-566
1-Thien-2-ylpropan-1-amine
1-Thien-2-ylpropan-1-amine

Hazard: Irritant

Formula: C₇H₁₁NS
MW: 141.24 g/mol
MDL Number: MFCD09040987
CAS Number: 6315-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR