4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid

Supplier: Matrix Scientific

037506-500MG
101843-556EA 109.25 USD
101843-556
4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid
4-(2-Oxo-2,3-dihydro-1H-indol-3-yl)butanoic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MW: 219.24 g/mol
MDL Number: MFCD06411619
CAS Number: 2971-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR