N-Butyl-2-chloro-N-methylacetamide

Supplier: Matrix Scientific

037504-500MG
101843-550EA 109.25 USD
101843-550
N-Butyl-2-chloro-N-methylacetamide
N-Butyl-2-chloro-N-methylacetamide

Hazard: Irritant

Formula: C₇H₁₄ClNO
MDL Number: MFCD03030385
CAS Number: 39096-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR