1-(Chloroacetyl)pyrrolidine

Supplier: Matrix Scientific

Synonyms: 2-Chloro-1-(pyrrolidin-1-yl)ethanone

037498-500MG
101843-538EA 146.15 USD
101843-538
1-(Chloroacetyl)pyrrolidine
1-(Chloroacetyl)pyrrolidine

Hazard: Irritant

Formula: C₆H₁₀ClNO
MW: 147.60 g/mol
MDL Number: MFCD01217237
CAS Number: 20266-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR