(2-Ethoxyphenyl)hydrazine hydrochloride

Supplier: Matrix Scientific

037497-500MG
101843-536EA 360 USD
101843-536
(2-Ethoxyphenyl)hydrazine hydrochloride
(2-Ethoxyphenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O
MDL Number: MFCD04966806
CAS Number: 126580-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR