5-(3,4-Dimethoxyphenyl)-1,3-oxazole-4-carboxylic acid

Supplier: Matrix Scientific

037490-500MG
101843-518EA 144.9 USD
101843-518
5-(3,4-Dimethoxyphenyl)-1,3-oxazole-4-carboxylic acid
5-(3,4-Dimethoxyphenyl)-1,3-oxazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₁NO₅
MDL Number: MFCD07376165
CAS Number: 89205-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR