5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-(4-Methoxyphenyl)oxazole-4-carboxylic acid

037489-500MG
101843-516EA 193.85 USD
101843-516
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉NO₄
MW: 219.2 g/mol
MDL Number: MFCD03644147
CAS Number: 89205-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR