(2E)-3-(3-Methoxyphenyl)acrylic acid

Synonyms: (E)-3-(3-Methoxyphenyl)acrylic acid

037483-500MG
101843-502EA 301.86 USD
101843-502
(2E)-3-(3-Methoxyphenyl)acrylic acid
(2E)-3-(3-Methoxyphenyl)acrylic acid

Hazard: Irritant

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD00004386
CAS Number: 17570-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR