trans-4-(Aminomethyl)cyclohexanecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: HAKUTAMCHAAMCAAMCHATranexamic acid

037476-25G
101843-482EA 29.23 USD
101843-482
trans-4-(Aminomethyl)cyclohexanecarboxylic acid ≥95%
trans-4-(Aminomethyl)cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD00001466
CAS Number: 1197-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR