1-Isoquinolin-5-ylmethanamine dihydrochloride

Supplier: Matrix Scientific

Synonyms: (Isoquinolin-5-yl)methanamine

037474-500MG
101843-472EA 300 USD
101843-472
1-Isoquinolin-5-ylmethanamine dihydrochloride
1-Isoquinolin-5-ylmethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂Cl₂N₂
MW: 231.13 g/mol
MDL Number: MFCD19442199
CAS Number: 58123-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR