(2,3-Dimethylphenyl)hydrazine

Supplier: Matrix Scientific

037462-500MG
101843-448EA 146.15 USD
101843-448
(2,3-Dimethylphenyl)hydrazine
(2,3-Dimethylphenyl)hydrazine

Hazard: Irritant

Formula: C₈H₁₂N₂
MDL Number: MFCD02656651
CAS Number: 84401-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR