1-(4-Methoxyphenyl)cyclohexanecarboxylic acid

Supplier: Matrix Scientific

037456-500MG
101843-428EA 146.15 USD
101843-428
1-(4-Methoxyphenyl)cyclohexanecarboxylic acid
1-(4-Methoxyphenyl)cyclohexanecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MDL Number: MFCD00001462
CAS Number: 7469-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR