Piperidin-4-one hydrochloride ≥97%

037452-25G 037452-5G
101843-414EA 43.08 USD
101843-414 101843-416
Piperidin-4-one hydrochloride ≥97%
Piperidin-4-one hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClNO
Melting Pt: 145…147 °C
MDL Number: MFCD00041019
CAS Number: 41979-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR