1-(4-Ethoxyphenyl)butan-1-one

Supplier: Matrix Scientific

037435-500MG
101843-368EA 146.15 USD
101843-368
1-(4-Ethoxyphenyl)butan-1-one
1-(4-Ethoxyphenyl)butan-1-one

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD11105547
CAS Number: 35031-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR