1-(4-Cyclohexylphenyl)butan-1-one

Supplier: Matrix Scientific

037432-500MG
101843-362EA 109.25 USD
101843-362
1-(4-Cyclohexylphenyl)butan-1-one
1-(4-Cyclohexylphenyl)butan-1-one

Hazard: Irritant

Formula: C₁₆H₂₂O
MDL Number: MFCD01098600
CAS Number: 70211-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR