2,2,3,3,4,4,5,5-Octafluoropentanal

Supplier: Matrix Scientific

Synonyms: 5H-Octafluoropentanal

037427-500MG
101843-350EA 146.15 USD
101843-350
2,2,3,3,4,4,5,5-Octafluoropentanal
2,2,3,3,4,4,5,5-Octafluoropentanal

Hazard: Irritant

Formula: C₅H₂F₈O
MW: 230.06 g/mol
MDL Number: MFCD00155897
CAS Number: 2648-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR