5-(4-Bromophenyl)-1,3,4-thiadiazol-2-amine

Supplier: Matrix Scientific

037426-500MG
101843-348EA 109.25 USD
101843-348
5-(4-Bromophenyl)-1,3,4-thiadiazol-2-amine
5-(4-Bromophenyl)-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₈H₆BrN₃S
MDL Number: MFCD00469706
CAS Number: 13178-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR