1-(4-Methoxyphenyl)cyclopentanecarbonitrile

037423-500MG
101843-342EA 401.25 USD
101843-342
1-(4-Methoxyphenyl)cyclopentanecarbonitrile
1-(4-Methoxyphenyl)cyclopentanecarbonitrile

Hazard: Irritant

Formula: C₁₃H₁₅NO
MDL Number: MFCD00065276
CAS Number: 1206-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR