3-(3-Methylphenyl)isoxazol-5-amine

Supplier: Matrix Scientific

037421-500MG
101843-338EA 96.6 USD
101843-338
3-(3-Methylphenyl)isoxazol-5-amine
3-(3-Methylphenyl)isoxazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MDL Number: MFCD06199358
CAS Number: 86685-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR