3-(4-Methoxyphenyl)isoxazol-5-amine

Supplier: Matrix Scientific

037420-500MG
101843-336EA 144.9 USD
101843-336
3-(4-Methoxyphenyl)isoxazol-5-amine
3-(4-Methoxyphenyl)isoxazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD03407397
CAS Number: 86685-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR