(4-Ethoxyphenyl)hydrazine hydrochloride

Supplier: Matrix Scientific

037414-500MG
101843-322EA 146.15 USD
101843-322
(4-Ethoxyphenyl)hydrazine hydrochloride
(4-Ethoxyphenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂O
MDL Number: MFCD12403348
CAS Number: 76014-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR