1-(4-Methoxyphenyl)cyclohexanecarbonitrile

Supplier: Matrix Scientific

037410-500MG
101843-308EA 109.25 USD
101843-308
1-(4-Methoxyphenyl)cyclohexanecarbonitrile
1-(4-Methoxyphenyl)cyclohexanecarbonitrile

Hazard: Irritant

Formula: C₁₄H₁₇NO
MDL Number: MFCD00021282
CAS Number: 36263-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR