(2E)-3-(3,4-Dimethoxyphenyl)acrylonitrile

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxycinnamonitrile

037409-500MG
101843-306EA 146.15 USD
101843-306
(2E)-3-(3,4-Dimethoxyphenyl)acrylonitrile
(2E)-3-(3,4-Dimethoxyphenyl)acrylonitrile

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MW: 189.22 g/mol
MDL Number: MFCD00001931
CAS Number: 6443-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR