1-(3-Cyanopropyl)pyrrolidine

Supplier: Matrix Scientific

Synonyms: 1-Pyrrolidinebutyronitrile1-Pyrrolidinebutanenitrile4-Pyrrolidinobutyronitrile

037408-500MG
101843-304EA 146.15 USD
101843-304
1-(3-Cyanopropyl)pyrrolidine
1-(3-Cyanopropyl)pyrrolidine

Hazard: Irritant

Formula: C₈H₁₄N₂
MW: 138.21 g/mol
MDL Number: MFCD00003183
CAS Number: 35543-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR