3-(6-Methylpyridin-2-yl)propan-1-ol

Supplier: Matrix Scientific

037405-500MG
101843-298EA 140 USD
101843-298
3-(6-Methylpyridin-2-yl)propan-1-ol
3-(6-Methylpyridin-2-yl)propan-1-ol

Hazard: Irritant

Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00012344
CAS Number: 61744-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR