1-Allylpyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

037396-500MG
101843-274EA 217.35 USD
101843-274
1-Allylpyrrolidine-2-carboxylic acid
1-Allylpyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₁₃NO₂
MDL Number: MFCD09026175
CAS Number: 678988-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR