5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

037393-500MG
101843-268EA 144.9 USD
101843-268
5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-amine
5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₈H₆FN₃O
Melting Pt: 251…253 °C
MDL Number: MFCD02073839
CAS Number: 7659-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR