2-Bromo-5-methyl-1,3,4-oxadiazole

Supplier: Matrix Scientific

037392-500MG
101843-266EA 362.25 USD
101843-266
2-Bromo-5-methyl-1,3,4-oxadiazole
2-Bromo-5-methyl-1,3,4-oxadiazole

Hazard: Irritant/Refrigerate

Formula: C₃H₃BrN₂O
MDL Number: MFCD12027126
CAS Number: 864750-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR