(4-Methyl-1H-benzimidazol-2-yl)methylamine hydrochloride

Supplier: Matrix Scientific

037389-500MG
101843-260EA 484.62 USD
101843-260
(4-Methyl-1H-benzimidazol-2-yl)methylamine hydrochloride
(4-Methyl-1H-benzimidazol-2-yl)methylamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClN₃
MDL Number: MFCD12027124
CAS Number: 933756-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR