1-(4-Methyl-1H-benzimidazol-2-yl)ethanol

Supplier: Matrix Scientific

037388-500MG
101843-258EA 146.15 USD
101843-258
1-(4-Methyl-1H-benzimidazol-2-yl)ethanol
1-(4-Methyl-1H-benzimidazol-2-yl)ethanol

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD09880340
CAS Number: 953857-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR