N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine hydrochloride

Supplier: Matrix Scientific

Synonyms: 1-Methyl-2-[(methylamino)methyl]-1H-benzimidazole

037387-500MG
101843-256EA 387.69 USD
101843-256
N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine hydrochloride
N-Methyl-N-[(1-methyl-1H-benzimidazol-2-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃
MW: 211.7 g/mol
MDL Number: MFCD12027123
CAS Number: 137898-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR