1-(5-Methyl-1H-benzimidazol-2-yl)ethanol

Supplier: Matrix Scientific

037385-500MG
101843-252EA 290.77 USD
101843-252
1-(5-Methyl-1H-benzimidazol-2-yl)ethanol
1-(5-Methyl-1H-benzimidazol-2-yl)ethanol

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MDL Number: MFCD00159954
CAS Number: 20033-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR