4-(Azidomethyl)benzamide

Supplier: Matrix Scientific

037382-500MG
101843-246EA 190.9 USD
101843-246
4-(Azidomethyl)benzamide
4-(Azidomethyl)benzamide

Hazard: Irritant

Formula: C₈H₈N₄O
MDL Number: MFCD12027121
CAS Number: 864528-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR