5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine

Supplier: Matrix Scientific

037379-500MG
101843-240EA 290.77 USD
101843-240
5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine
5-(3-Fluorophenyl)-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₈H₆FN₃O
MDL Number: MFCD09028454
CAS Number: 341978-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR