Pyrazine-2-carbohydrazide

Supplier: Matrix Scientific

Synonyms: Pyrazinoic acid hydrazide

037378-500MG
101843-238EA 290.77 USD
101843-238
Pyrazine-2-carbohydrazide
Pyrazine-2-carbohydrazide

Hazard: Irritant

Formula: C₅H₆N₄O
MW: 138.13 g/mol
MDL Number: MFCD00778762
CAS Number: 768-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR