5-Pyrazin-2-yl-1,3,4-oxadiazol-2-amine
Supplier: Matrix Scientific
037377-500MG
101843-236EA
290.77
USD
101843-236
5-Pyrazin-2-yl-1,3,4-oxadiazol-2-amine
5-Pyrazin-2-yl-1,3,4-oxadiazol-2-amine
Hazard: Irritant
Formula:
C₆H₅N₅O |
MDL Number:
MFCD05722375 CAS Number: 717848-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...