1,3-Benzoxazol-2-ylmethylamine hydrochloride

Supplier: Matrix Scientific

037376-500MG
101843-234EA 493.85 USD
101843-234
1,3-Benzoxazol-2-ylmethylamine hydrochloride
1,3-Benzoxazol-2-ylmethylamine hydrochloride

Hazard: Irritant

Formula: C₈H₉ClN₂O
MDL Number: MFCD09763577
CAS Number: 1072806-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR