Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyanobenzohydrazide hydrochloride

Supplier: Matrix Scientific


037367-500MG
101843-216EA 162.15 USD InStock
101843-216
4-Cyanobenzohydrazide hydrochloride
4-Cyanobenzohydrazide hydrochloride
CAS: 1865223-60-4
MDL Number: MFCD30181469
Molecular Formula: C8H8ClN3O
Molecular Weight: 197,63
Hazard: Irritant


Formula: C₈H₈ClN₃O
MW: 197,63 g/mol
MDL Number: MFCD30181469
CAS Number: 1865223-60-4
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty